missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FRS2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 11503-1-AP-20UL
Informations supplémentaires : Peso : 0.00010kg
Description
Adapter protein that links activated FGR and NGF receptors to downstream signaling pathways. Plays an important role in the activation of MAP kinases and in the phosphorylation of PIK3R1, the regulatory subunit of phosphatidylinositol 3- kinase, in response to ligand-mediated activation of FGFR1. Modulates signaling via SHC1 by competing for a common binding site on NTRK1.Spécification
FRS2 | |
Polyclonal | |
Unconjugated | |
FRS2 | |
FGFR signaling adaptor SNT, FGFR substrate 2, FRS2, FRS2A, FRS2alpha, SNT, SNT 1, SNT1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10818, 314850, 327826 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry | |
0.22 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8C180, Q8WU20 | |
Frs2 | |
FRS2 Fusion Protein Ag2052 | |
20 μL | |
Primary | |
Human, Rat, Mouse | |
Antibody | |
IgG |