missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GAC Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 19958-1-AP-20UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
This antibody is specific to GAC isoform. It has no cross reaction to KGA.
Sahai (1983) demonstrated phosphate-activated glutaminase (EC 3.5.1.2) in human platelets. It is the major enzyme yielding glutamate from glutamine. Significance of the enzyme derives from its possible implication in behavior disturbances in which glutamate acts as a neurotransmitter(Prusiner, 1981). High heritability of platelet glutaminase was indicated by studies of Sahai and Vogel (1983) who found an intraclass correlation coefficient of 0.96 for monozygotic twins and 0.53 for dizygotic twins.Specifica
GAC | |
Polyclonal | |
Unconjugated | |
GLS | |
AAD20, GAC, GAC-specific, GLS, GLS1, glutaminase C, K glutaminase, KIAA0838, L glutamine amidohydrolase | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
14660, 24398, 2744 | |
-20°C | |
Liquid |
Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
0.29 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
D3Z7P3, O94925, P13264 | |
GLS | |
Peptide | |
20 μL | |
Primary | |
Human, Rat, Mouse | |
Antibody | |
IgG |