missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gastrokine 1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 19344-1-AP-20UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
Gastrokine 1, also known as GKN1 and CA11, is a 185 amino acid containing secreted protein belonging to the gastrokine family with a BRICHOS domain and an asn-rich region. GKN1 (also known as an antioncogene) is reported to be down-regulated in human gastric cancer tissue as compared to normal gastric mucosa. Differential display of normal and cancerous gastric tissue suggested that this protein is predomitly expressed in stomach and the expression is down-regulated to considerable frequency in cancer tissue or gastric cancer cell lines examined. This extra cellular protein is co-secreted with mucins and is a signaling growth factor with mitogenic activity and may be involved in maintaining the integrity of the gastric mucosal epithelium.Specifica
Gastrokine 1 | |
Polyclonal | |
Unconjugated | |
GKN1 | |
18 kDa antrum mucosa protein, AMP 18, AMP18, BRICD1, CA11, FOV, foveolin, gastrokine 1, GKN1, Protein CA11 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
56287, 66283 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.43 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9CR36, Q9NS71 | |
GKN1 | |
Gastrokine 1 Fusion Protein Ag6091 | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |