missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gelsolin Mouse anti-Human, Clone: 3C4A2, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 66280-1-IG-20UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
The protein encoded by this gene binds to the "plus" ends of actin monomers and filaments to prevent monomer exchange. The encoded calcium-regulated protein functions in both assembly and disassembly of actin filaments. Defects in this gene are a cause of familial amyloidosis Finnish type (FAF). Multiple transcript variants encoding several different isoforms have been found for this gene.Specifica
Gelsolin | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P06396 | |
GSN | |
Gelsolin Fusion Protein Ag17916 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgA |
Western Blot, Immunohistochemistry (Paraffin) | |
3C4A2 | |
Unconjugated | |
GSN | |
Actin depolymerizing factor, ADF, AGEL, Brevin, Gelsolin, GSN | |
Mouse | |
Affinity Chromatography | |
RUO | |
2934 | |
-20°C | |
Liquid |