missing translation for 'onlineSavingsMsg'
Learn More

Gelsolin Mouse anti-Human, Clone: 3C4A2, Proteintech

Mouse Monoclonal Antibody

Marca:  Proteintech 66280-1-IG-150UL

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16898005

  • 325.38€ / 150µL

Spedizione stimata: 11-10-2022
per visualizzare lo stock disponibile
Aggiungi al carrello

Descrizione

Descrizione

The protein encoded by this gene binds to the "plus" ends of actin monomers and filaments to prevent monomer exchange. The encoded calcium-regulated protein functions in both assembly and disassembly of actin filaments. Defects in this gene are a cause of familial amyloidosis Finnish type (FAF). Multiple transcript variants encoding several different isoforms have been found for this gene.
Specifica

Specifica

Gelsolin
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P06396
GSN
Gelsolin Fusion Protein Ag17916
150 μL
Primary
Human
Antibody
IgA
Western Blot, Immunohistochemistry (Paraffin)
3C4A2
Unconjugated
GSN
Actin depolymerizing factor, ADF, AGEL, Brevin, Gelsolin, GSN
Mouse
Affinity Chromatography
RUO
2934
-20°C
Liquid
Videos
SDS
Documenti
Certificati
Offerte speciali

Offerte speciali