Learn More
Thermo Scientific™ GENESYS Fiber Optic Probes
Dip, tap and measure - eliminate liquid transfer and cuvettes from your workflow.
Marca: Thermo Scientific™ 840-307400
Dettagli aggiuntivi : Peso : 0.00500kg
Descrizione
Dip, tap and measure - eliminate liquid transfer and cuvettes from your workflow.
Preserve sample integrity while minimizing sample carryover.Specifica
10 mm Pathlength Tip for Dip Probe for GENESYS 150 and GENESYS 180 Spectrophotometers | |
Tip for Dip Probe | |
GENESYS 150 and GENESYS 180 spectrophotometers | |
10 mm |