Learn More
Gilson™ PIPETMAN™ EXPERT™ Sterilized Filter Pipette Tips
PIPETMAN™ EXPERT™ Sterilized Filter Pipette Tips features graduations at varying volumes allowing for quick-checks when aspirating samples.
Marca: Gilson™ F1732032
Dettagli aggiuntivi : Peso : 6.55000kg
Descrizione
- Features patented, low-retention polymer that creates hydrophobic surface to minimize sample binding when dispensing.
- Color-coded tip packaging corresponds to industry standards for volume color matching to pipettes.
- Free of Human DNA, RNase, DNase, PCR Inhibitors and Pyrogens.
Specifica
0.1 to 10 μL | |
Sterile | |
Autoclavable | |
4.534 cm | |
Racked | |
Red | |
Yes | |
Polypropylene | |
2 μL, 5 μL, 10 μL |
Filtered | |
Clear | |
1.78 in. | |
Rack | |
Pipette Tips | |
Single-use | |
Pipetman EXPERT | |
Certified Quality |