Learn More
Hirschmann™ Class A USP Volumetric Flasks
Allows for precise, safe liquid measurement and handling in the laboratory. Hirschmann Instruments™ Class A USP Volumetric Flasks are manufactured from durable DURAN™ glass tubing and are available with a Batch Certificate of Performance, an Individual USP-Certificate of Performance, and a DAkkS-Certificate.
Marca: Hirschmann™ 282018927
Dettagli aggiuntivi : Peso : 0.25000kg
Descrizione
- Blue graduations
- With dated batch identification
- With NS ground and polyethylene stopper
Specifica
USP | |
500 mL | |
260 ± 5 mm | |
0.15 mL | |
A |
Polyethylene | |
Borosilicate Glass | |
19/26 NS Standard Ground | |
Yes |