missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GOLGA2/GM130 Mouse anti-Human, Rat, Clone: 2A4F11, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 66662-1-IG-150UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
The Golgi apparatus, which participates in glycosylation and transport of proteins and lipids in the secretory pathway, consists of a series of stacked cisternae (flattened membrane sacs). Interactions between the Golgi and microtubules are thought to be important for the reorganization of the Golgi after it fragments during mitosis. This gene encodes one of the golgins, a family of proteins localized to the Golgi. This encoded protein has been postulated to play roles in the stacking of Golgi cisternae and in vesicular transport. Several alternatively spliced transcript variants of this gene have been described, but the full-length nature of these variants has not been determined.Specifica
GOLGA2/GM130 | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q08379, Q62839 | |
GOLGA2 | |
GOLGA2/GM130 Fusion Protein Ag1848 | |
150 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG1 |
Western Blot | |
2A4F11 | |
Unconjugated | |
GOLGA2 | |
GM130, GM130 autoantigen, GOLGA2, Golgin 95, Golgin subfamily A member 2 | |
Mouse | |
Protein G | |
RUO | |
2801, 64528 | |
-20°C | |
Liquid |