Learn More
Fisherbrand™ Carta filtro tecnica in cellulosa di grado 1103
Suitable for removal of fine particles
Marca: Fisherbrand™
Dettagli aggiuntivi : Peso : 0.01000kg
Descrizione
- Thick smooth paper
- Wet strengthened
Specifica
Disc | |
12 to 16 μm | |
For dyes, Ether, Alcoholic solutions, Eaux de Cologne, salted solutions, vegetal extracts, hair solutions, perfumes, tinctures, saccharine juices, vinegar | |
100 Pack | |
240 mm |
Cellulose | |
12 to 16 μm | |
1103 | |
0.3 mm | |
240 mm |