Learn More
Fisherbrand™ Grade 1107 Cellulose Technical Filter Paper: Discs
Used for filtration of semi fine particles
Marca: Fisherbrand™
Dettagli aggiuntivi : Peso : 0.01000kg
Descrizione
- Thick smooth paper with medium pore structure
Specifica
Folded Disc | |
20 to 30 μm | |
For Serum, Culture Medium, dyes, tinctures, vegetal extracts, turbid liquids, juices, Eaux de Cologne, perfumes, essences, hair tonics | |
100 Pack | |
500 mm |
Cellulose | |
20 to 30 μm | |
1107 | |
0.38 mm | |
500 mm |