Learn More
Sartorius™ Grade 470 Folded Filter Cellulose and Diatomaceous Earth Filter Papers
Offers better separating capability and quickly retains the finest particles at high flow rates.
Marca: Sartorius™ FT-4-606-125
Dettagli aggiuntivi : Peso : 0.10000kg
Descrizione
- Used for the filtration of the finest, semi-colloidal precipitates, e.g. those consisting of proteins, clay or cold-precipitated barium
- For clarification of beer, wine, urine during spectophotometric or refractometric test
Specifica
Folded | |
80sec. | |
470 | |
0.32 mm | |
125 mm |
Cellulose, Diatomaceous Earth | |
Offer a much better separating capability than pure cellulose papers at the same rate of filtration | |
100/Pk. | |
Diatomaceous Earth Filter Paper |