Learn More
Sartorius Stedim Biotech™ Grade 6 S/N Creped Papers for Qualitative and Technical Analyses, Folded
Sartorius Stedim Biotech™ Grade 6 S/N Creped Papers for Qualitative and Technical Analyses, Folded are suitable for analytical and technical purposes and feature a fast flow rate due to their structure.
Marca:  Sartorius Stedim Biotech™ FT-4-314-125
Dettagli aggiuntivi : Peso : 0.17800kg
Codice prodotto. 15992990
  • 230.00€ / pezzi 100
Spedizione stimata: 11-06-2024
per visualizzare lo stock disponibile
Aggiungi al carrello
Descrizione
Descrizione
  • Made of refined pulp and cotton linters with > 95% alpha-cellulose
  • Ash content between 0.1 and 0.15%
  • Medium fast filtering
  • Wet-strengthened
  • Non-sterile, bulk-packed
Specifica
Specifica
145 g/m2
Cotton Linters, Refined Pulp
12 sec.
100/Pk.
Qualitative Filter Paper
Folded
6 to 14 μm
6 S/N
0.55 mm
125 mm