Learn More
Sartorius™ Grade 6 Smooth Surface Folded Filter Qualitative Filter Papers
Used for analytical and technical purposes.
Marca: Sartorius™ FT-3-312-070
Dettagli aggiuntivi : Peso : 0.10000kg
Descrizione
Used for analytical and technical purposesSpecifica
Disc | |
15μm/sec. | |
6 | |
0.17 mm | |
70 mm |
Cotton Linters, Refined Pulp | |
For qualitative-technical analyses, used for routine analyses like clarification, determination of substances, but also as discs with a center hole for technical applications | |
100 Pack | |
Qualitative Filter Paper | |
70 mm |