missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GRIM19 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 10986-1-AP-20UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
Mitochondrial NADH ubiquinone oxidoreductase (complex I) catalyzes the first step in the electron transport chain, the oxidation of NADH to NAD+ coupled to proton translocation across the inner mitochondrial membrane. GRIM-19 is a component of the complex I and is a product of a cell death regulatory gene induced by interferon-b and retinoic acid. GRIM-19 has previously been detected in HeLa cells, predomitly in the nucleus, though punctate staining of the cytoplasm was also observed.Specifica
GRIM19 | |
Polyclonal | |
Unconjugated | |
NDUFA13 | |
B16.6, CDA016, CGI 39, CI B16.6, Complex I B16.6, GRIM 19, GRIM19, NDUFA13 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
100911483, 51079, 67184 | |
-20°C | |
Liquid |
Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunoprecipitation, Immunofluorescence | |
0.21 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9ERS2, Q9P0J0 | |
NDUFA13 | |
GRIM19 Fusion Protein Ag1435 | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |