missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GSDMD Mouse anti-Human, Clone: 2E6C7, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 66387-1-IG-150UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
FOXN3 is a member of the forkhead/winged helixtranscription factor family. Checkpoints are eukaryotic DNA damage-inducible cell cycle arrests at G1 and G2. Check point suppressor 1 suppresses multiple yeast checkpoint mutationsincluding mec1, rad9, rad53 and dun1 by activating aMEC1-independent checkpoint pathway. Alternative splicing isobserved at the locus, resulting in distinct isoforms.Specifica
GSDMD | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P57764 | |
GSDMD | |
GSDMD Fusion Protein Ag16827 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG2a |
Immunocytochemistry, Immunofluorescence, Western Blot | |
2E6C7 | |
Unconjugated | |
GSDMD | |
DF5L, DFNA5L, FKSG10, gasdermin D, GSDMD, GSDMDC1 | |
Mouse | |
Protein A | |
RUO | |
79792 | |
-20°C | |
Liquid |