missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GSTA4 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 17271-1-AP-150UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
Conjugation of reduced glutathione to a wide number of exogenous and endogenous hydrophobic electrophiles. This isozyme has a high catalytic efficiency with 4-hydroxyalkenals such as 4-hydroxynonenal (4-HNE).Specifica
GSTA4 | |
Polyclonal | |
Unconjugated | |
GSTA4 | |
Glutathione S transferase A4, Glutathione S transferase A4 4, GST class alpha member 4, GSTA4, GSTA4 4, GTA4 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
2941 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
0.28 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O15217 | |
GSTA4 | |
GSTA4 Fusion Protein Ag11028 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |