missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HAT1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 11432-1-AP-20UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
The protein encoded by this gene is a type B histone acetyltransferase (HAT) that is involved in the rapid acetylation of newly synthesized cytoplasmic histones, which are in turn imported into the nucleus for de novo deposition onto nascent DNA chains. Histone acetylation, particularly of histone H4, plays an important role in replication-dependent chromatin assembly. Specifically, this HAT can acetylate soluble but not nucleosomal histone H4 at lysines 5 and 12, and to a lesser degree, histone H2A at lysine 5. Alternatively spliced transcript variants have been identified for this gene.Specifica
HAT1 | |
Polyclonal | |
Unconjugated | |
HAT1 | |
HAT1, histone acetyltransferase 1, KAT1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
107435, 296501, 8520 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O14929, Q5M939, Q8BY71 | |
HAT1 | |
HAT1 Fusion Protein Ag1952 | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |