Learn More
Memmert™ HEPA14-Filter For Chamber
HEPA Filter sterile, individually packed, with fixing unit
Marca: Memmert™ ICO150B49800
Dettagli aggiuntivi : Peso : 0.01100kg
Descrizione
- Includes fixing unit
- According to EN 1822
- Packed in sterile condition
Specifica
ICO150 |