Learn More
Memmert™ HEPA14-Filter For Chamber
HEPA Filter sterile, individually packed, with fixing unit
Marca: Memmert™ ICO50B49800
Dettagli aggiuntivi : Peso : 0.01100kg
Descrizione
- Includes fixing unit
- According to EN 1822
- Packed in sterile condition
Specifica
Individually packed | |
ICO50 | |
Fixing unit |