missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hephaestin Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 11148-1-AP-150UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
B1AJX8(HEPH) is similar to an iron transport protein found in mouse. The mouse protein is similar to ceruloplasmin, a serum multi-copper ferroxidase, and is thought to be a membrane-bound protein responsible for transport of dietary iron from epithelial cells of the intestinal lumen into the circulatory system. In mouse, defects in this gene can lead to severe microcytic anemia. Three transcript variants encoding different isoforms have been described for this gene. The protein encoded by this gene is similar to an iron transport protein found in mouse. The mouse protein is similar to ceruloplasmin, a serum multi-copper ferroxidase, and is thought to be a membrane-bound protein responsible for transport of dietary iron from epithelial cells of the intestinal lumen into the circulatory system. In mouse, defects in this gene can lead to severe microcytic anemia. Two transcript variants encoding different isoforms have been described for this gene.Specifica
Hephaestin | |
Polyclonal | |
Unconjugated | |
HEPH | |
CPL, HEPH, Hephaestin, KIAA0698 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
117240, 15203, 9843 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.17 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q920H8, Q9BQS7, Q9Z0Z4 | |
HEPH | |
Hephaestin Fusion Protein Ag1604 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |