missing translation for 'onlineSavingsMsg'
Learn More

HESX1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marca:  Proteintech 17927-1-AP-20UL

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16897934

  • 121.38€ / 20µL

Spedizione stimata: 05-10-2022
per visualizzare lo stock disponibile
Aggiungi al carrello

Descrizione

Descrizione

This gene encodes a conserved homeobox protein that is a transcriptional repressor in the developing forebrain and pituitary gland. Mutations in this gene are associated with septooptic dysplasia, HESX1-related growth hormone deficiency, and combined pituitary hormone deficiency.
Specifica

Specifica

HESX1
Polyclonal
Unconjugated
HESX1
ANF, hAnf, HESX homeobox 1, HESX1, Homeobox protein ANF, RPX
Rabbit
Antigen Affinity Chromatography
RUO
15209, 498575, 8820
-20°C
Liquid
Immunohistochemistry (Paraffin)
0.24 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q61658, Q9UBX0
Hesx1
HESX1 Fusion Protein Ag12348
20 μL
Primary
Mouse, Rat, Human
Antibody
IgG
Videos
SDS
Documenti
Certificati
Offerte speciali

Offerte speciali