Learn More
HESX1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 17927-1-AP-150UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
This gene encodes a conserved homeobox protein that is a transcriptional repressor in the developing forebrain and pituitary gland. Mutations in this gene are associated with septooptic dysplasia, HESX1-related growth hormone deficiency, and combined pituitary hormone deficiency.Specifica
HESX1 | |
Polyclonal | |
Unconjugated | |
HESX1 | |
ANF, hAnf, HESX homeobox 1, HESX1, Homeobox protein ANF, RPX | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
15209, 498575, 8820 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.24 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q61658, Q9UBX0 | |
Hesx1 | |
HESX1 Fusion Protein Ag12348 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |