Learn More
Emma Safety Footwear Himalaya Safety Shoes
A modern shoe that not only looks good but also protects your feet from top to bottom and everywhere else. Himalaya features a steel toe cap and an integrated stainless steel plate. The double-layer, anti-slip PU sole provides extra protection from hazards on the ground.
Marca: Emma Safety Footwear MM33454842
Dettagli aggiuntivi : Peso : 0.40000kg
Descrizione
- S3-Safety shoe with sole plate for wet work
- Oil Nubuck construction
- GripForce™ Easy Twist
- 3M™ Reflective Striping
- Hydro-Tec™ Sanitized Silver for drier, fresher feet
- Sanitized
Specifica
EN ISO 20345:2011 | |
Black | |
Hydro-Tec™ Sanitized Silver | |
42 |
Laces | |
Himalaya | |
Oiled Nubuck | |
Steel |