missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HMCN1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 18837-1-AP-20UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
This antibody raised against a synthesized peptide corresponding to 960-916 amino acids of human HMCN1 can recognize all three isoforms of HMCN1.
Hemicentin-1 (HMCN1), also known as fibulin-6 (FIBL-6), is an extracellular protein of the immunoglobulin superfamily. HMCN1 consists of a von Willebrand A domain, tandem immunoglobulin modules, tandem EGF modules, and a fibulin carboxy-terminal module. Mutations of HMCN1 gene may be associated with age-related macular degeneration. Three alternative transcripts encoding different HMCN1 isoforms have been described.Specifica
HMCN1 | |
Polyclonal | |
Unconjugated | |
HMCN1 | |
ARMD1, FBLN6, FIBL 6, FIBL6, Fibulin 6, hemicentin 1, HMCN1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
289094, 545370, 83872 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.71 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
D3YXG0, Q96RW7 | |
Hmcn1 | |
Peptide | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG |