Learn More
Hoefer™ SUB13 Midi Submarine Combs
Accessories for SUB13 Midi Submarine
Marca: Hoefer™ SUB13-C2-20
Dettagli aggiuntivi : Peso : 0.01000kg
Descrizione
- Ranges from 10, 12, 16, 20, 24 and 28 Wells
- 1, 1.5 and 2 mm Thickness
- 3 to 9.5 mm Well Width
- 13 min., 85 max. μL Sample Volume
Specifica
SUB13 Midi Submarine Gel Electrophoresis Unit | |
20 | |
0.2 cm | |
0.15 in. | |
0.19 in. |
Comb | |
0.07 in. | |
35 μL | |
0.4 cm | |
0.5 cm |