Learn More
Hoefer™ SUB20 Maxi-Standard and SUB20C Maxi-Cooled Submarine Combs
Accessories for SUB20 Maxi-Standard and SUB20C Maxi-Cooled Submarines
Marca: Hoefer™ SUB20-C1-16
Dettagli aggiuntivi : Peso : 0.01000kg
Descrizione
- Ranges from 16, 20, 28 and 40 Wells
- 1, 1.5 and 2 mm Thickness
- 3, 5, 7 and 8.5 mm Well Width
- Min. 13, max. 75 μL Sample Volume
Specifica
SUB20 Maxi Standard and SUB20C Maxi Cooled Submarine | |
16 | |
0.1 cm | |
0.33 in. | |
0.19 in. |
Comb | |
0.03 in. | |
35 μL | |
0.85 cm | |
0.5 cm |