Learn More
DWK Life Sciences Holding Device for Reaction Vessels, from chrome-nickel steel
The holding device is attached to a tripod linkage and serves to suspend and hold reaction vessels within the fume hood.
Marca: DWK Life Sciences 290734602
850.25 EUR valido fino al 2024-12-31
Usa il codice promo "23258" per avere il tuo prezzo promozionale.
Dettagli aggiuntivi : Peso : 1.63700kg
Descrizione
Reaction vessels can be operated and tempered in various ways. To prevent the reaction apparatus from moving during the test, the reaction vessel should ideally be suspended from a tripod linkage by means of a holding device. Cooling and heating media can thus be fed flexibly from below to the reaction vessel and removed again.
- For fixed suspension of the lid and the reaction vessel on a tripod rod
Specifica
DN 100 | |
Holder for lids and reaction vessels; Mounting | |
No |
Chrome-Nickel Steel | |
1/Pk. |