missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Holding Device for Reaction Vessels, from chrome-nickel steel
The holding device is attached to a tripod linkage and serves to suspend and hold reaction vessels within the fume hood.
895.00€ - 935.00€
Specifica
Materiale | Chrome-Nickel Steel |
---|---|
Da utilizzare con (applicazione) | Holder for lids and reaction vessels; Mounting |
N. per confezione | 1/Pk. |
Autoclavabile | No |
Codice del prodotto | Marca | Descrizione | Prezzo | Quantità e disponibilità | |||||
---|---|---|---|---|---|---|---|---|---|
Codice del prodotto | Marca | Descrizione | Prezzo | Quantità e disponibilità | |||||
10675401
|
DWK Life Sciences
290735701 |
DN 150 |
935.00€
Pezzo |
Spedizione stimata: 05-06-2024
Eseguire il login per visualizzare lo stock disponibile |
|||||
10665401
|
DWK Life Sciences
290734602 |
DN 100 |
895.00€
Pezzo |
Spedizione stimata: 23-05-2024
Eseguire il login per visualizzare lo stock disponibile |
|||||
Descrizione
Reaction vessels can be operated and tempered in various ways. To prevent the reaction apparatus from moving during the test, the reaction vessel should ideally be suspended from a tripod linkage by means of a holding device. Cooling and heating media can thus be fed flexibly from below to the reaction vessel and removed again.
- For fixed suspension of the lid and the reaction vessel on a tripod rod
Specifica
Chrome-Nickel Steel | |
Holder for lids and reaction vessels; Mounting | |
1/Pk. | |
No |