Learn More
Honeywell™ Mont-Blanc S3 Safety Shoes
Marca: Honeywell™ 6233215-37/7
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
European Directive 89.686 CEE, EN 345-1, ISO 9001:2008 | |
For food services, pharmaceutical and medical use | |
37 | |
Double density Polyurethane | |
Metallic | |
Safety Shoe |
White | |
Leather | |
Anti-puncture Steel midsole, white color, made up of double density Polyurethane | |
Slip-on safety | |
Impact, splash, slip-resistant |