Learn More
Honeywell™ Pro'Clean S2 Shoes
Marca: Honeywell™ 6246127-39/7
Dettagli aggiuntivi : Peso : 0.87300kg
Specifica
EN ISO 20345: 2011, SRC Certified | |
For professional use such as catering, industrial cleaning, laboratory, medical and pharmaceutical | |
39 | |
Double density Polyurethane | |
Abrasion, fuel oil and grease | |
Shoes |
White | |
Microfiber | |
Removable, anti-static insole | |
Spacium non-metallic | |
Yes |