Learn More
Abnova™ Human ANXA13 Full-length ORF (NP_001003954.1, 1 a.a. - 357 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00000312-P01.25ug
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
This gene encodes a member of the annexin family. Members of this calcium-dependent phospholipid-binding protein family play a role in the regulation of cellular growth and in signal transduction pathways. The specific function of this gene has not yet been determined; however, it is associated with the plasma membrane of undifferentiated, proliferating endothelial cells and differentiated villus enterocytes. Alternatively spliced transcript variants encoding different isoforms have been identified. [provided by RefSeq]
Sequence: MGNRHSQSYTLSEGSQQLPKGDSQPSTVVQPLSHPSRNGEPEAPQPAKASSPQGFDVDRDAKKLNKACKGMGTNEAAIIEILSGRTSDERQQIKQKYKATYGKELEEVLKSELSGNFEKTALALLDRPSEYAARQLQKAMKGLGTDESVLIEVLCTRTNKEIIAIKEAYQRLFDRSLESDVKGDTSGNLKKILVSLLQANRNEGDDVDKDLAGQDAKDLYDAGEGRWGTDELAFNEVLAKRSYKQLRATFQAYQILIGKDIEEAIEEETSGDLQKAYLTLVRCAQDCEDYFAERLYKSMKGAGTDEETLIRIVVTRAEVDLQGIKAKFQEKYQKSLSDMVRSDTSGDFRKLLVALLHSpecifica
NP_001003954.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
66.1kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
RUO | |
ANXA13 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
312 | |
ANXA13 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ANX13/ISA/MGC150460 | |
ANXA13 | |
Recombinant | |
wheat germ expression system |