Learn More
Abnova™ Human CARS Partial ORF (NP_001742, 447 a.a. - 546 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00000833-Q01.10ug
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
This gene encodes a class 1 aminoacyl-tRNA synthetase, cysteinyl-tRNA synthetase. Each of the twenty aminoacyl-tRNA synthetases catalyzes the aminoacylation of a specific tRNA or tRNA isoaccepting family with the cognate amino acid. This gene is one of several located near the imprinted gene domain of 11p15.5, an important tumor-suppressor gene region. Alterations in this region have been associated with the Beckwith-Wiedemann syndrome, Wilms tumor, rhabdomyosarcoma, adrenocortical carcinoma, and lung, ovarian, and breast cancer. Alternative splicing of this gene results in multiple transcript variants encoding distinct isoforms. [provided by RefSeq]
Sequence: NTMESALQYEKFLNEFFLNVKDILRAPVDITGQFEKWGEEEAELNKNFYDKKTAIHKALCDNVDTRTVMEEMRALVSQCNLYMAARKAVRKRPNQALLENSpecifica
NP_001742 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
NTMESALQYEKFLNEFFLNVKDILRAPVDITGQFEKWGEEEAELNKNFYDKKTAIHKALCDNVDTRTVMEEMRALVSQCNLYMAARKAVRKRPNQALLEN | |
RUO | |
CARS | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
833 | |
CARS (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CARS1/CYSRS/MGC:11246 | |
CARS | |
Recombinant | |
wheat germ expression system |