Learn More
ACROBiosystems Human CEACAM-5 Protein, Fc Tag
Marca:  ACROBiosystems CE5-H5253-100ug
Dettagli aggiuntivi : Peso : 0.00010kg
Codice prodotto. 17226170
  • 391.93€ / 100µg
Spedizione stimata: 30-04-2024
per visualizzare lo stock disponibile
Descrizione
Descrizione
Carcinoembryonic antigen-related cell adhesion molecule 5 (CEACAM5) is also Carcinoembryonic antigen (CEA), Meconium antigen 100, CD antigen CD66e, CEACAM5 belongs to the immunoglobulin superfamily and CEA family. CEACAM5 contains seven Ig-like (immunoglobulin-like) domains. CEACAM5 is homodimer protein which binding of E.coli Dr adhesins leads to dissociation of the homodimer. CEACAM5 is cell surface glycoprotein that plays a role in cell adhesion and in intracellular signaling. CEACAM5 is receptor for E.coli Dr adhesins.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 97.8 kDa.
Specifica
Specifica
97.8 kDa
Human CEACAM-5, Fc Tag (CE5-H5253) is expressed from human 293 cells (HEK293). It contains AA Lys 35 - Ala 685 (Accession # P06731-1).
-20°C
CEACAM-5
Human
HEK293
Bio-Markers & CD Antigens
100 μg
Lys 35 - Ala 685
CEACAM-5,CD66e,CEA,Meconium antigen 100
Unconjugated
Fc Tag
Powder