missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human CLGN Partial ORF (NP_004353.1, 21 a.a. - 120 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00001047-Q01.25ug
Dettagli aggiuntivi : Peso : 0.02000kg
Descrizione
Calmegin is a testis-specific endoplasmic reticulum chaperone protein. CLGN may play a role in spermatogeneisis and infertility. [provided by RefSeq]
Sequence: FMDDDVETEDFEENSEEIDVNESELSSEIKYKTPQPIGEVYFAETFDSGRLAGWVLSKAKKDDMDEEISIYDGRWEIEELKENQVPGDRGLVLKSRAKHHSpecifica
NP_004353.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
FMDDDVETEDFEENSEEIDVNESELSSEIKYKTPQPIGEVYFAETFDSGRLAGWVLSKAKKDDMDEEISIYDGRWEIEELKENQVPGDRGLVLKSRAKHH | |
RUO | |
CLGN | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
1047 | |
CLGN (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CLGN | |
Wheat Germ (in vitro) | |
GST | |
Liquid |