Learn More
Abnova™ Human CTNNBL1 Partial ORF (AAH36739, 210 a.a. - 311 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00056259-Q01.10ug
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
The protein encoded by this gene contains an acidic domain, a putative bipartite nuclear localization signal, a nuclear export signal, a leucine-isoleucine zipper, and phosphorylation motifs. In addition, the encoded protein contains Armadillo/beta-catenin-like repeats, which have been implicated in protein-protein interactions. Although the function of this protein has not been determined, the C-terminal portion of the protein has been shown to possess apoptosis-inducing activity. [provided by RefSeq]
Sequence: EKHDMVRRGEIIDNDTEEEFYLRRLDAGLFVLQHICYIMAEICNANVPQIRQRVHQILNMRGSSIKIVRHIIKEYAENIGDGRSPEFRENEQKRILGLLENFSpecifica
AAH36739 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.85kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
EKHDMVRRGEIIDNDTEEEFYLRRLDAGLFVLQHICYIMAEICNANVPQIRQRVHQILNMRGSSIKIVRHIIKEYAENIGDGRSPEFRENEQKRILGLLENF | |
RUO | |
CTNNBL1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
56259 | |
CTNNBL1 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
C20orf33/FLJ21108/NAP/NYD-SP19/P14L/PP8304/dJ633O20.1 | |
CTNNBL1 | |
Recombinant | |
wheat germ expression system |