Learn More
Abnova™ Human HOXB3 Partial ORF (NP_002137.4, 272 a.a. - 370 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00003213-Q01.25ug
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
This gene is a member of the Antp homeobox family and encodes a nuclear protein with a homeobox DNA-binding domain. It is included in a cluster of homeobox B genes located on chromosome 17. The encoded protein functions as a sequence-specific transcription factor that is involved in development. Increased expression of this gene is associated with a distinct biologic subset of acute myeloid leukemia (AML). [provided by RefSeq]
Sequence: TAGFMNALHSMTPSYESPSPPAFGKAHQNAYALPSNYQPPLKGCGAPQKYPPTPAPEYEPHVLQANGGAYGTPTMQGSPVYVGGGGYADPLPPPAGPSLSpecifica
NP_002137.4 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
TAGFMNALHSMTPSYESPSPPAFGKAHQNAYALPSNYQPPLKGCGAPQKYPPTPAPEYEPHVLQANGGAYGTPTMQGSPVYVGGGGYADPLPPPAGPSL | |
RUO | |
HOXB3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
3213 | |
HOXB3 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
HOX2/HOX2G/Hox-2.7 | |
HOXB3 | |
Recombinant | |
wheat germ expression system |