missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human KLRG1 Partial ORF (NP_005801, 57 a.a. - 119 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00010219-Q01.25ug

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16172166

  • 528.00€ / 25µg
Spedizione stimata: 03-11-2023
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

Natural killer (NK) cells are lymphocytes that can mediate lysis of certain tumor cells and virus-infected cells without previous activation. They can also regulate specific humoral and cell-mediated immunity. The protein encoded by this gene belongs to the killer cell lectin-like receptor (KLR) family, which is a group of transmembrane proteins preferentially expressed in NK cells. Studies in mice suggested that the expression of this gene may be regulated by MHC class I molecules. Alternatively spliced transcript variants have been reported, but their full-length natures have not yet been determined. [provided by RefSeq]

Sequence: QWILCQGSNYSTCASCPSCPDRWMKYGNHCYYFSVEEKDWNSSLEFCLARDSHLLVITDNQEM
Specifica

Specifica

NP_005801
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
32.67kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
QWILCQGSNYSTCASCPSCPDRWMKYGNHCYYFSVEEKDWNSSLEFCLARDSHLLVITDNQEM
RUO
KLRG1
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
10219
KLRG1 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
2F1/CLEC15A/MAFA/MAFA-2F1/MAFA-L/MAFA-LIKE/MGC13600
KLRG1
Recombinant
wheat germ expression system
Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali