Learn More
Abnova™ Human LGALS14 Partial ORF (NP_064514.1, 73 a.a. - 139 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00056891-Q01.10ug
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
This gene is predominantly expressed in placenta. The encoded protein belongs to the galectin (galaptin/S-lectin) family. The members of galectin family contain one or two carbohydrate recognition domains, which can bind beta-galactoside. Two alternatively spliced transcript variants encoding distinct isoforms have been observed. [provided by RefSeq]
Sequence: RYEEKCYYLPFEDGKPFELCIYVRHKEYKVMVNGQRIYNFAHRFPPASVKMLQVLRDISLTRVLISDSpecifica
NP_064514.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
33.11kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
RYEEKCYYLPFEDGKPFELCIYVRHKEYKVMVNGQRIYNFAHRFPPASVKMLQVLRDISLTRVLISD | |
RUO | |
LGALS14 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
56891 | |
LGALS14 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CLC2/MGC22235/PPL13 | |
LGALS14 | |
Recombinant | |
wheat germ expression system |