Learn More
ACROBiosystems Human Mucin-1 / MUC-1 (54-115) Protein, Fc Tag
Marca: ACROBiosystems MU1-H5257-100ug
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
Membrane mucins have several functions in epithelial cells including cytoprotection, extravasation during metastases, maintenance of luminal structure, and signal transduction. MUC17, contains an extended, repetitive extracellular glycosylation domain and a carboxyl terminus with two EGF-like domains, a SEA module domain, a transmembrane domain, and a cytoplasmic domain with potential serine and tyrosine phosphorylation sites. Interacts via its C-terminus with PDZK1 and this interaction appears important for proper localization. Probably plays a role in maintaining homeostasis on mucosal surfaces.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 33.6 kDa.Specifica
NP_002447.4 | |
100 µg | |
Leu 54 - Gly 115 | |
Mucin 1,MUC1,CD227,EMA,H23AG,KL-6,MAM6,MUC-1/SEC,MUC-1/X,MUC1/ZD,PEM,PEMT,PUM,CA 15-3,Episialin | |
Unconjugated | |
Fc Tag | |
HEK293 | |
Bio-Markers and CD Antigens |
33.6 kDa | |
Human Mucin-1 (54-115), Fc Tag (MU1-H5257) is expressed from human 293 cells (HEK293). It contains AA Leu 54 - Gly 115 (Accession # NP_002447.4). | |
-20°C | |
Mucin-1 | |
Human | |
Less than 1.0 EU / µg by the LAL method. | |
Powder |