Learn More
Abnova™ Human NFE2L3 Partial ORF (NP_004280, 585 a.a. - 694 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00009603-Q01.25ug
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
This gene encodes a member of the cap 'n' collar basic-region leucine zipper family of transcription factors. The encoded protein heterodimerizes with small musculoaponeurotic fibrosarcoma factors to bind antioxidant response elements in target genes. This protein is a membrane bound glycoprotein that is targeted to the endoplasmic reticulum and the nuclear envelope. Pseudogenes of this gene are found on chromosomes 16, 17, and 18. [provided by RefSeq]
Sequence: RGKNKVAAQNCRKRKLDIILNLEDDVCNLQAKKETLKREQAQCNKAINIMKQKLHDLYHDIFSRLRDDQGRPVNPNHYALQCTHDGSILIVPKELVASGHKKETQKGKRKSpecifica
NP_004280 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.84kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
RGKNKVAAQNCRKRKLDIILNLEDDVCNLQAKKETLKREQAQCNKAINIMKQKLHDLYHDIFSRLRDDQGRPVNPNHYALQCTHDGSILIVPKELVASGHKKETQKGKRK | |
RUO | |
NFE2L3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
9603 | |
NFE2L3 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
NRF3 | |
NFE2L3 | |
Recombinant | |
wheat germ expression system |