missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human OTOF Partial ORF (NP_919224.1, 1 a.a. - 98 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00009381-Q01.10ug

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16160526

  • 335.00€ / 10µg
Spedizione stimata:
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

Mutations in this gene are a cause of neurosensory nonsyndromic recessive deafness, DFNB9. The short form of the encoded protein has 3 C2 domains, a single carboxy-terminal transmembrane domain found also in the C. elegans spermatogenesis factor FER-1 and human dysferlin, while the long form has 6 C2 domains. The homology suggests that this protein may be involved in vesicle membrane fusion. Several transcript variants encoding multiple isoforms have been found for this gene. [provided by RefSeq]

Sequence: MALLIHLKTVSELRGRGDRIAKVTFRGQSFYSRVLENCEDVADFDETFRWPVASSIDRNEMLEIQVFNYSKVFSNKLIGTFRMVLQKVVEESHVEVTD
Specifica

Specifica

NP_919224.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.52kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
MALLIHLKTVSELRGRGDRIAKVTFRGQSFYSRVLENCEDVADFDETFRWPVASSIDRNEMLEIQVFNYSKVFSNKLIGTFRMVLQKVVEESHVEVTD
RUO
OTOF
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
9381
OTOF (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DFNB6/DFNB9/FER1L2/NSRD9
OTOF
Recombinant
wheat germ expression system
Suggerimenti di prodotto

Suggerimenti di prodotto

Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali