Learn More
Abnova™ Human OTOF Partial ORF (NP_919224.1, 1 a.a. - 98 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00009381-Q01.25ug
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
Mutations in this gene are a cause of neurosensory nonsyndromic recessive deafness, DFNB9. The short form of the encoded protein has 3 C2 domains, a single carboxy-terminal transmembrane domain found also in the C. elegans spermatogenesis factor FER-1 and human dysferlin, while the long form has 6 C2 domains. The homology suggests that this protein may be involved in vesicle membrane fusion. Several transcript variants encoding multiple isoforms have been found for this gene. [provided by RefSeq]
Sequence: MALLIHLKTVSELRGRGDRIAKVTFRGQSFYSRVLENCEDVADFDETFRWPVASSIDRNEMLEIQVFNYSKVFSNKLIGTFRMVLQKVVEESHVEVTDSpecifica
NP_919224.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.52kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MALLIHLKTVSELRGRGDRIAKVTFRGQSFYSRVLENCEDVADFDETFRWPVASSIDRNEMLEIQVFNYSKVFSNKLIGTFRMVLQKVVEESHVEVTD | |
RUO | |
OTOF | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
9381 | |
OTOF (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DFNB6/DFNB9/FER1L2/NSRD9 | |
OTOF | |
Recombinant | |
wheat germ expression system |