Learn More
ACROBiosystems Human PD-1 / PDCD1 Full Length Protein, His Tag
Marca:  ACROBiosystems PD1-H52H6-20ug
Dettagli aggiuntivi : Peso : 0.00010kg
 vedi altre versioni di questo prodotto
Codice prodotto. 17226410
  • dietro preventivo / 20µg
Questo articolo richiede un aggiornamento del prezzo prima di essere ordinato.
Richiedi qui il prezzo per questo articolo.
Descrizione
Descrizione
Programmed cell death protein 1 (PD-1) is also known as CD279 and PDCD1, is a type I membrane protein and is a member of the extended CD28/CTLA-4 family of T cell regulators. PDCD1 is expressed on the surface of activated T cells, B cells, macrophages, myeloid cells and a subset of thymocytes. PD-1 has two ligands, PD-L1 and PD-L2, which are members of the B7 family. PD-L1 is expressed on almost all murine tumor cell lines, including PA1 myeloma, P815 mastocytoma, and B16 melanoma upon treatment with IFN-γ. PD-L2 expression is more restricted and is expressed mainly by DCs and a few tumor lines. PD1 inhibits the T-cell proliferation and production of related cytokines including IL-1, IL-4, IL-10 and IFN-γ by suppressing the activation and transduction of PI3K/AKT pathway. In addition, coligation of PD1 inhibits BCR-mediating signal by dephosphorylating key signal transducer. In vitro, treatment of anti-CD3 stimulated T cells with PD-L1-Ig results in reduced T cell proliferation and IFN-γ secretion. Monoclonal antibodies targeting PD-1 that boost the immune system are being developed for the treatment of cancer.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 30.7 kDa. The protein migrates as 40-120 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Specifica
Specifica
NP_005009.2
>80% as determined by SDS-PAGE.
Human PD-1 Full Length, His Tag (PD1-H52H6) is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Leu 288 (Accession # Q15116-1).
-20°C
PD-1
His Tag
Less than 1.0 EU / µg by the LAL method.
Powder
Human
30.7 kDa
20 µg
Leu 25 - Leu 288
PDCD1,PD1,CD279,SLEB2
Unconjugated
50 mM Tris, 100 mM NaCl, pH8.0
HEK293
Immune Checkpoint Proteins