Learn More
Abnova™ Human PGBD1 Partial ORF (NP_115896.1, 241 a.a. - 350 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00084547-Q01.10ug
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
The piggyBac family of proteins, found in diverse animals, are transposases related to the transposase of the canonical piggyBac transposon from the moth, Trichoplusia ni. This family also includes genes in several genomes, including human, that appear to have been derived from the piggyBac transposons. This gene belongs to the subfamily of piggyBac transposable element derived (PGBD) genes. The PGBD proteins appear to be novel, with no obvious relationship to other transposases, or other known protein families. This gene product is specifically expressed in the brain, however, its exact function is not known. [provided by RefSeq]
Sequence: NLCGNSAQETVMSLSPMTEEIVTKDRLFKAKQETSEEMEQSGEASGKPNRECAPQIPCSTPIATERTVAHLNTLKDRHPGDLWARMHISSLEYAAGDITRKGRKKDKARVSpecifica
NP_115896.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.84kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
NLCGNSAQETVMSLSPMTEEIVTKDRLFKAKQETSEEMEQSGEASGKPNRECAPQIPCSTPIATERTVAHLNTLKDRHPGDLWARMHISSLEYAAGDITRKGRKKDKARV | |
RUO | |
PGBD1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
84547 | |
PGBD1 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
HUCEP-4/SCAND4/dJ874C20.4 | |
PGBD1 | |
Recombinant | |
wheat germ expression system |