Learn More
ACROBiosystems Human PTK7 / CCK4 Protein, His Tag (MALS verified)
Marca: ACROBiosystems PT7-H52H3-1mg
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
PTK7, highly expressed in lung, liver, pancreas, kidney, placenta and melanocytes. Weakly expressed in thyroid gland, ovary, brain, heart and skeletal muscle. Also expressed in erythroleukemia cells. Inactive tyrosine kinase involved in Wnt signaling pathway. Component of both the non-canonical (also known as the Wnt/planar cell polarity signaling) and the canonical Wnt signaling pathway. Functions in cell adhesion, cell migration, cell polarity, proliferation, actin cytoskeleton reorganization and apoptosis. Has a role in embryogenesis, epithelial tissue organization and angiogenesis.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 76.5 kDa. The protein migrates as 90-115 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Specifica
NP_002812.2 | |
1 mg | |
Ala 31 - Thr 704 | |
PTK7,CCK4,CCK-4,Tyrosine-protein kinase-like 7,Protein-tyrosine kinase 7,Protein-tyrosine kinase 7 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE. |
76.5 kDa | |
Human PTK7, His Tag (PT7-H52H3) is expressed from human 293 cells (HEK293). It contains AA Ala 31 - Thr 704 (Accession # Q13308-1). | |
-20°C | |
PTK7 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Enzymes |