Learn More
Abnova™ Human SLC39A8 Full-length ORF (NP_071437.3, 1 a.a. - 460 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00064116-P01.10ug
Descrizione
This gene encodes a member of the SLC39 family of solute-carrier genes, which show structural characteristics of zinc transporters. The encoded protein is glycosylated and found in the plasma membrane and mitochondria, and functions in the cellular import of zinc at the onset of inflammation. It is also thought to be the primary transporter of the toxic cation cadmium, which is found in cigarette smoke. Multiple transcript variants encoding different isoforms have been found for this gene. Additional alternatively spliced transcript variants of this gene have been described, but their full-length nature is not known. [provided by RefSeq]
Sequence: MAPGRAVAGLLLLAAAGLGGVAEGPGLAFSEDVLSVFGANLSLSAAQLQHLLEQMGAASRVGVPEPGQLHFNQCLTAEEIFSLHGFSNATQITSSKFSVICPAVLQQLNFHPCEDRPKHKTRPSHSEVWGYGFLSVTIINLASLLGLILTPLIKKSYFPKILTFFVGLAIGTLFSNAIFQLIPEAFGFDPKVDSYVEKAVAVFGGFYLLFFFERMLKMLLKTYGQNGHTHFGNDNFGPQEKTHQPKALPAINGVTCYANPAVTEANGHIHFDNVSVVSLQDGKKEPSSCTCLKGPKLSEIGTIAWMITLCDALHNFIDGLAIGASCTLSLLQGLSTSIAILCEEFPHELGDFVILLNAGMSTRQALLFNFLSACSCYVGLAFGILVGNNFAPNIIFALAGGMFLYISLADMFPEMNDMLREKVTGRKTDFTFFMIQNAGMLTGFTAILLITLYAGEIELESpecifica
NP_071437.3 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
76kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 μg | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
BIGM103/LZT-Hs6/PP3105/ZIP8 | |
SLC39A8 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
64116 | |
SLC39A8 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MAPGRAVAGLLLLAAAGLGGVAEGPGLAFSEDVLSVFGANLSLSAAQLQHLLEQMGAASRVGVPEPGQLHFNQCLTAEEIFSLHGFSNATQITSSKFSVICPAVLQQLNFHPCEDRPKHKTRPSHSEVWGYGFLSVTIINLASLLGLILTPLIKKSYFPKILTFFVGLAIGTLFSNAIFQLIPEAFGFDPKVDSYVEKAVAVFGGFYLLFFFERMLKMLLKTYGQNGHTHFGNDNFGPQEKTHQPKALPAINGVTCYANPAVTEANGHIHFDNVSVVSLQDGKKEPSSCTCLKGPKLSEIGTIAWMITLCDALHNFIDGLAIGASCTLSLLQGLSTSIAILCEEFPHELGDFVILLNAGMSTRQALLFNFLSACSCYVGLAFGILVGNNFAPNIIFALAGGMFLYISLADMFPEMNDMLREKVTGRKTDFTFFMIQNAGMLTGFTAILLITLYAGEIELE | |
RUO | |
SLC39A8 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.