Learn More
Abnova™ Human USP30 Partial ORF (NP_116052.1, 401 a.a. - 508 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00084749-Q01.25ug
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
USP30, a member of the ubiquitin-specific protease family (see USP1, MIM 603478), is a novel mitochondrial deubiquitinating (DUB) enzyme (Nakamura and Hirose, 2008 [PubMed 18287522]).[supplied by OMIM]
Sequence: SLLPTLSAPMPFPLPVVPDYSSSTYLFRLMAVVVHHGDMHSGHFVTYRRSPPSARNPLSTSNQWLWVSDDTVRKASLQEVLSSSAYLLFYERVLSRMQHQSQECKSEESpecifica
NP_116052.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.62kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
SLLPTLSAPMPFPLPVVPDYSSSTYLFRLMAVVVHHGDMHSGHFVTYRRSPPSARNPLSTSNQWLWVSDDTVRKASLQEVLSSSAYLLFYERVLSRMQHQSQECKSEE | |
RUO | |
USP30 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
84749 | |
USP30 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ40511/MGC10702 | |
USP30 | |
Recombinant | |
wheat germ expression system |