missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human ZNF44 Full-length ORF (AAH32246.1, 1 a.a. - 154 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00051710-P01.10ug
Dettagli aggiuntivi : Peso : 0.00010kg
Description
Sequence: MALCYGTFWGYPKMLEAANLMEGLVDIGPWVTLPRGQPEVLEWGLPKDQDSVAFEDVAVNFTHEEWALLGPSQKNLYRDVMRETIRNLNCIGMKWENQNIDDQHQNLRRNPRLSETWLQNFILIHYGHLVALSDGGLLQFSTGQGLPVTQAGVQSpécification
AAH32246.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
43.9kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp434F1811/DKFZp686L21136/GIOT-2/KOX7/ZNF/ZNF504/ZNF55/ZNF58 | |
ZNF44 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
51710 | |
ZNF44 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MALCYGTFWGYPKMLEAANLMEGLVDIGPWVTLPRGQPEVLEWGLPKDQDSVAFEDVAVNFTHEEWALLGPSQKNLYRDVMRETIRNLNCIGMKWENQNIDDQHQNLRRNPRLSETWLQNFILIHYGHLVALSDGGLLQFSTGQGLPVTQAGVQ | |
RUO | |
ZNF44 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |