Learn More
Corning™ Stripettor™ Ultra Pipet Controller Accessory, Hydrophobic PTFE Filters
Extreme chemical resistance of PTFE membranes makes them very useful for filtering solvents or other aggressive chemicals for which other membranes are unsuitable.
Marca: Corning™ 4992
Dettagli aggiuntivi : Peso : 0.20000kg
Allerta:
Because of their hydrophobicity, PTFE membranes must be prewetted with a solvent, such as ethanol, before aqueous solutions can be filtered.
Descrizione
These filters are use in the Corning™ StripettorTM Ultra Pipet Controller.
Specifica
White | |
Hydrophobic PTFE Filter | |
Stripettor™ Ultra Pipet Controller |
0.45 μm | |
Polytetrafluorethylene (PTFE) | |
5/Pk. |